Learn More
Description
Specifications
Specifications
| Antigen | Sars Envelope Protein |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, ELISA 1-3 ug/ml |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P59637 |
| Gene Alias | SARS coronavirus Envelope protein, SARS CoV E protein, SARS CoV Envelope protein, SARSCoV, SARSCoV Envelope protein, Severe acute respiratory syndrome |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
