Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sars Membrane Protein Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241059MFV500
Description
Sars Membrane Protein Polyclonal antibody specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for ELISASpecifications
Sars Membrane Protein | |
Polyclonal | |
ELISA | |
E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to a 13 aa peptide from within the first 50 amino acids from the M (Membrane protein) for the human SARS coronavirus (Genbank accession no. AFR58746.1) Amino Acid Squence: ADNGTITVEELKQ | |
0.1 mL | |
Immunology, Virology Bacteria and Parasites | |
1489672 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
SARS-CoV | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction