Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Nucleocapsid Protein Antibody (AP201054) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP290967
Description
SARS Nucleocapsid Protein Monoclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay.Specifications
SARS Nucleocapsid Protein | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with No Preservative | |
Mouse | |
Protein G purified | |
RUO | |
1489678 | |
SARS-CoV, SARS-CoV-2 | |
Lyophilized |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay | |
AP201054 | |
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence, Immunoassay, Direct ELISA | |
COVID-19 nucleocapsid protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARSCOV2 N protein, SARS-COV-2 nucleocapsid protein, SARS-COV-2 Nucleoprotein, Severe acute respiratory syndrome | |
The antibody was developed by immunizing mice with a with a protein fragment corresponding to amino acids 1-49 (C-MSDNGPQSNQRSAPRITFGGPTDSTDNNQNGGRNGARPKQRRPQGLPNN) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. NP_828858.1) | |
100 μg | |
Primary | |
Reconstitute with sterilized PBS to a final concentration is 0.5 mg/ml. Aliquot and store frozen at <−20 for at least for six months without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction