Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Nucleocapsid Protein Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056576PE
Description
SARS Nucleocapsid Protein Polyclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV, SARS-CoV-2, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Dual RNAscope ISH-IHCSpecifications
SARS Nucleocapsid Protein | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page, Dual RNAscope ISH-IHC | |
N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome | |
The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2) | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
1489678 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
SARS-CoV, SARS-CoV-2, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction