Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Monoclonal Antibody (14B3D), Bioss
SDP

Catalog No. 502281183
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1183 Supplier Bioss Supplier No. BSM49134M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS Nucleocapsid Protein Monoclonal antibody specifically detects SARS Nucleocapsid Protein in Virus samples. It is validated for ELISA, Western Blot

Specifications

Antigen SARS Nucleocapsid Protein
Applications ELISA, Western Blot
Classification Monoclonal
Clone 14B3D
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% ProClin 300
Gene Alias Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
Host Species Mouse
Immunogen Recombinant SARS N protein (no tag)
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.