Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS RDRP Antibody (4E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | SARS RDRP |
---|---|
Clone | 4E6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000 |
Applications | Western Blot |
Description
SARS RDRP Monoclonal antibody specifically detects SARS RDRP in SARS-CoV samples. It is validated for Western BlotSpecifications
SARS RDRP | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS | |
IgG1 | |
Protein A purified |
4E6 | |
Western Blot 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
SARS-CoV | |
The antibody was developed by immunizing mice with a GST fusion protein corresponding to Replicase polyprotein 1ab for the Human SARS coronavirus (Genbank accession no. NP_828849.2) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title