Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS/SARS-CoV-2 Spike Protein S2 Monoclonal Antibody (GT8110)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA542339
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The S2 domain plays a role in fusion. Further, S proteins plays an important role in the induction of neutralizing-antibodies and T-cell responses, as well as protective immunity. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
Specifications
| SARS/SARS-CoV-2 Spike Protein S2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| GT8110 | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 S2; COVID19 Spike S2; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| Recombinant protein encompassing a sequence within the extracellular domain of SARS-CoV-2 (COVID-19) Spike S2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction