Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Spike Protein Antibody (16F1071R), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP290998B
Description
SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Sars Spike Protein | |
Monoclonal | |
Biotin | |
E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
This recombinant antibody was developed by immunizing mice with a synthetic peptide corresponding to amino acids 19-35 (CTTFDDVQAPNYTQHTS) from the S (Spike glycoprotein)(Spike Protein 1) for the Human SARS coronavirus (Genbank accession no. NP_828851.1) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunofluorescence | |
16F1071R | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
SARS-CoV | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction