Learn More
Specifications
Specifications
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56685 |
| Gene Alias | E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 |
| Quantity | 0.1 mg |
| Research Category | Core ESC Like Genes, Immunology, Stem Cell Markers, Virology Bacteria and Parasites |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
