Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Spike Protein Antibody (D005), Alexa Fluor™ 350, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP290989AF350
Description
SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA, Product Image.Specifications
Sars Spike Protein | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein SARS-CoV-2 Omicron (B.1.1.529) Spike S1 Protein SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 Protein SARS-CoV Spike S1 Protein SARS-CoV-2 Spike S1 Protein SARS-CoV-2 Spike RBD Protein No cross-reactivity in ELISA with: MERS-CoV Spike S1 Protein HCoV-HKU1 (isolate N1) Spike S1 Protein HCoV-HKU1 (isolate N5) Spike S1 Protein HCoV-NL63 Spike S1 Protein HCoV-229E Spike S1 Protein HCoV-OC43 Spike S1+S2 ECD Protein | |
Store at 4°C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
D005 | |
ELISA | |
E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
Recombinant SARS-CoV Spike RBD Protein | |
0.1 mL | |
Primary | |
SARS-CoV-2, SARS-CoV | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction