Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056684 0.2 mL
NB10056684T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056684 Supplier Novus Biologicals Supplier No. NB10056684
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS Spike Protein Polyclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot.

Specifications

Antigen Sars Spike Protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. NP_006504.2
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 553-570 (C-TESNKKFLPFQQFGRDIA from the S (Spike glycoprotein)(Spike protein 1) for the Human SARS coronavirus (Genbank accession no. YP_009724390)
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489668
Target Species SARS-CoV
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.