Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Spike Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Sars Spike Protein |
---|---|
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056684T
![]() |
Novus Biologicals
NB10056684SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056684
![]() |
Novus Biologicals
NB10056684 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
SARS Spike Protein Polyclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot.Specifications
Sars Spike Protein | |
Western Blot | |
Unconjugated | |
Rabbit | |
SARS-CoV | |
NP_006504.2 | |
1489668 | |
IgG | |
Unpurified |
Western Blot 1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 553-570 (C-TESNKKFLPFQQFGRDIA from the S (Spike glycoprotein)(Spike protein 1) for the Human SARS coronavirus (Genbank accession no. YP_009724390) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title