Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Sars Spike Protein
Dilution Western Blot 1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056685T
SDP
View Documents
Novus Biologicals
NB10056685SS
0.05 mL
Each for $181.90
Only null left
Add to Cart
 
NB10056685
SDP
View Documents
Novus Biologicals
NB10056685
0.2 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

SARS Spike Protein Polyclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot.
Specifications

Specifications

Sars Spike Protein
Western Blot
Unconjugated
Rabbit
SARS-CoV
NP_006504.2
1489668
IgG
Unpurified
Western Blot 1:1000
Polyclonal
Antisera
RUO
Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide
E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 441-453 (C-LDSKVGGNYNYLY) from the S (Spike glycoprotein)(Spike protein 1) for the Human SARS coronavirus (Genbank accession no. YP_009724390)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.