Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS Coronavirus Matrix Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572456
Description
SARS Coronavirus Matrix Polyclonal Antibody for ELISA
A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. The M protein (Membrane protein, Matrix protein) is one of the major structural viral proteins. It is an integral membrane protein involved in the budding of the viral particles and interacts with S (Spike) protein and the nucleocapsid protein.
Specifications
SARS Coronavirus Matrix | |
Polyclonal | |
Unconjugated | |
M | |
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Matrix Protein; SARS-CoV2; SARS-CoV-2; SARS-M | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
1489672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P59596 | |
M | |
KLH conjugated synthetic peptide between 192~221 amino acids from the C-terminus region of SARS M protein. | |
400 μL | |
Primary | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction