Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531473 Supplier Invitrogen™ Supplier No. PA531473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (84%), Rat (84%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SARS2
Gene Accession No. Q9NP81
Gene Alias 2410015F05Rik; D7Ertd353e; EC 6.1.1.11; mitochondrial seryl-tRNA synthetase; mtSerRS; SARS; Sars2; Sarsm; serine tRNA ligase 2, mitochondrial; serine-tRNA ligase; serine-tRNA ligase, mitochondrial; serine--tRNA ligase, mitochondrial; SerRS; SerRS2; SerRSmt; SERS; seryl-aminoacyl-tRNA synthetase 2; Seryl-tRNA synthetase; seryl-tRNA synthetase 2; seryl-tRNA synthetase 2, mitochondrial; seryl-tRNA synthetase, mitochondrial; seryl-tRNA(Ser/Sec) synthetase; SYS; zgc:66345
Gene Symbols SARS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SARS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.