Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110341 Supplier Invitrogen™ Supplier No. PA5110341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARFKDLNLAG TAEVGLAGYF MDHTVAFRDL PVRMVCSSTC YRAETNTGQE PRGLYRVHHF TKVEMFGVTG PGLEQSSQLL EEFLSLQMEI LTELGLHFRV LDMPTQELGL PAYRKFDIEA WMPGRGRFGE VTSASNCTDF QSRRLHIMFQ TEAGELQFAH TVNATACAVP RLLIALLESN QQKDGSVLVP PALQSYLGTD RITAPTHVPL QYIGPNQPRK PGLPGQPAVS.

This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.046 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SARS2
Gene Accession No. Q9JJL8, Q9NP81
Gene Alias 2410015F05Rik; D7Ertd353e; EC 6.1.1.11; mitochondrial seryl-tRNA synthetase; mtSerRS; SARS; Sars2; Sarsm; serine tRNA ligase 2, mitochondrial; serine-tRNA ligase; serine-tRNA ligase, mitochondrial; serine--tRNA ligase, mitochondrial; SerRS; SerRS2; SerRSmt; SERS; seryl-aminoacyl-tRNA synthetase 2; Seryl-tRNA synthetase; seryl-tRNA synthetase 2; seryl-tRNA synthetase 2, mitochondrial; seryl-tRNA synthetase, mitochondrial; seryl-tRNA(Ser/Sec) synthetase; SYS; zgc:66345
Gene Symbols SARS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 289-518 of human SARS2 (NP_060297.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54938, 71984
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.