Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111361
Description
Immunogen sequence: QIYNIDPARF KDLNLAGTAE VGLAGYFMDH TVAFRDLPVR MVCSSTCYRA ETNTGQEPRG LYRVHHFTKV EMFGVTGPGL.
This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.
Specifications
| SARS2 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP81 | |
| SARS2 | |
| Recombinant Protein corresonding to Human SARS2. Recombinant protein control fragment (Product #RP-101647). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| SARS2 | |
| 2410015F05Rik; D7Ertd353e; EC 6.1.1.11; mitochondrial seryl-tRNA synthetase; mtSerRS; SARS; Sars2; Sarsm; serine tRNA ligase 2, mitochondrial; serine-tRNA ligase; serine-tRNA ligase, mitochondrial; serine--tRNA ligase, mitochondrial; SerRS; SerRS2; SerRSmt; SERS; seryl-aminoacyl-tRNA synthetase 2; Seryl-tRNA synthetase; seryl-tRNA synthetase 2; seryl-tRNA synthetase 2, mitochondrial; seryl-tRNA synthetase, mitochondrial; seryl-tRNA(Ser/Sec) synthetase; SYS; zgc:66345 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54938 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction