Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SART-3 Mouse anti-Mouse, Human, Clone: 1C4H6, MilliporeSigma™

Mouse Monoclonal Antibody

$481.79 - $485.79

Specifications

Antigen SART-3
Clone 1C4H6
Applications Depletion Assay, Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE190925
View Documents
MilliporeSigma
MABE190925UL
25 μL
Each for $485.79
Only null left
Add to Cart
 
MABE1909100
View Documents
MilliporeSigma
MABE1909100UL
100 μL
Each for $481.79
Only null left
Add to Cart
 
Description

Description

SART-3 Monoclonal antibody specifically detects SART-3 in Mouse, Human samples. It is validated for Immunodepletion, Immunofluorescence, Western Blotting.
Specifications

Specifications

SART-3
Depletion Assay, Immunofluorescence, Western Blot
Unconjugated
Mouse
Mouse, Human
GST-tagged full-length human Squamous cell carcinoma antigen recognized by T-cells 3 (SART-3).
Primary
Stable for 1 year at 2-8°C from date of receipt.
1C4H6
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.