Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SART1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214836JF549
Description
SART1 Polyclonal specifically detects SART1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SART1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Allergen Hom s 1, Ara1, HAF, HOMS1, hSART-1, hSnu66, hypoxia-associated factor, IgE autoantigen, MGC2038, SART-1, SART1(259) protein, SART1(800) protein, SART1259, small nuclear ribonucleoprotein 110kDa (U4/U6.U5), SNRNP110, Snu66, SNU66 homolog, squamous cell carcinoma antigen recognised by T cells, squamous cell carcinoma antigen recognized by T cells, Squamous cell carcinoma antigen recognized by T cells 1, U4/U6.U5 tri-snRNP-associated 110 kDa protein, U4/U6.U5 tri-snRNP-associated protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SART1 | |
A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290] | |
0.1 mL | |
Hypoxia | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction