Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SART1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597120 Supplier Invitrogen™ Supplier No. PA597120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSSKKHRGE KEAAGTTAAA GTGGATEQPP RHREHKKHKH RSGGSGGSGG ERRKRSRERG GERGSGRRGA EAEARSSTHG RERSQAEPSE RRVKREKRDD GYEAAASSKT SSGDASSLSI EETNKLRAKL GLKPLEVNAI KKEAGTKEEP VTADVINPMA LRQREELREK LAAAKEKRLL NQKLGKIKTL GEDDPWLDDT AAWIERSRQL QKEKDLAEKR AKLLEEMDQE FGVSTLVEEE FGQRRQDLYS ARDLQ; Positive Samples: HeLa, Jurkat, HepG2, Mouse liver, Mouse brain, Mouse testis, Rat brain; Cellular Location: Nucleus.

This gene encodes two proteins, the SART1protein expressed in the nucleus of the majority of proliferating cells, and the SART1protein expressed in the cytosol of epithelial cancers. The SART1protein is translated by the mechanism of -1 frameshifting during posttranscriptional regulation; its full-length sequence is not published yet. The two encoded proteins are thought to be involved in the regulation of proliferation. Both proteins have tumor-rejection antigens. The SART1protein possesses tumor epitopes capable of inducing HLA-A2402-restricted cytotoxic T lymphocytes in cancer patients. This SART1antigen may be useful in specific immunotherapy for cancer patients and may serve as a paradigmatic tool for the diagnosis and treatment of patients with atopy. The SART1protein is found to be essential for the recruitment of the tri-snRNP to the pre-spliceosome in the spliceosome assembly pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SART1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SART1
Gene Accession No. O43290, Q5XIW8, Q9Z315
Gene Alias Ara1; Haf; Hom s 1; HOMS1; hSART-1; hSnu66; Hypoxia-associated factor; IgE autoantigen; MGC2038; mSART-1; rSART-1; Sart1; SART-1; SART1(259) protein; SART1(800) protein; SART1259; small nuclear ribonucleoprotein 110kDa (U4/U6.U5); SNRNP110; Snu66; SNU66 homolog; squamous cell carcinoma antigen recognised by T cells; squamous cell carcinoma antigen recognized by T cells; squamous cell carcinoma antigen recognized by T cells 1; squamous cell carcinoma antigen recognized by T-cells 1; U4/U6.U5 tri-snRNP-associated 110 kDa protein; U4/U6.U5 tri-snRNP-associated protein 1; U5-110K
Gene Symbols SART1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human SART1 (NP_0051371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20227, 29678, 9092
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.