Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SART3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518120
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518120 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518120 Supplier Invitrogen™ Supplier No. PA518120
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SART3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SART3
Gene Accession No. Q15020
Gene Alias AU045857; DSAP1; HIV-1 Tat-interacting protein of 110kDa; hSART-3; Kiaa0156; MGC138188; mKIAA0156; mSART-3; P100; p110; p110 nuclear RNA-binding protein; p110(nrb); RP11-13G14; Sart3; SART-3; spliceosome associated factor 3, U4/U6 recycling protein; squamous cell carcinoma antigen recognised by T cells 3; squamous cell carcinoma antigen recognized by T cells 3; squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110; Tumor-rejection antigen SART3
Gene Symbols SART3
Host Species Goat
Immunogen Synthetic peptide sequence (MSNADFAKLFLRK) corresponding to the C-terminus amino acids of SART3 (aa 951-963).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9733
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.