Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SART3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528625

Catalog No. PIPA528625


Only null left
Add to Cart

Description

Description

Recommended positive controls: A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SART3
Polyclonal
Unconjugated
SART3
AU045857; DSAP1; HIV-1 Tat-interacting protein of 110kDa; hSART-3; Kiaa0156; MGC138188; mKIAA0156; mSART-3; P100; p110; p110 nuclear RNA-binding protein; p110(nrb); RP11-13G14; Sart3; SART-3; spliceosome associated factor 3, U4/U6 recycling protein; squamous cell carcinoma antigen recognised by T cells 3; squamous cell carcinoma antigen recognized by T cells 3; squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110; Tumor-rejection antigen SART3
Rabbit
Antigen affinity chromatography
RUO
9733
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
0.52 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15020
SART3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SART3. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.