Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Sartobind Lab Affinity IDA75

Catalog No. 14557115 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
2.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-557-115 2.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-557-115 Supplier Sartorius Supplier No. 93IDA42DB12V
Only null left
Add to Cart
Add to Cart

Ready-to-use membrane adsorbers for immobilized metal affinity chromatography (IMAC) in syringe filter format. Superior flow rates and high binding capacity based on the patented macroporous Sartobind™ membranes with iminodiacetic acid (IDA).

  • Based on the Patented Sartobind™ Platform Sartobind™ Lab membrane adsorbers for immobilized metal affinity chromatography (IMAC) are ready-to-use units in a syringe filter format.
  • They are based on the patented macroporous Sartobind™ membranes, with iminodiacetic acid (IDA) covalently bound to the entire internal surface of the pores. This results in a chromatography matrix with superior flow rates and high binding capacity.
  • The IDA ligand enables coupling of the metal ion of your choice to the membrane adsorber, ensuring simple process optimization for maximum purity and yield of each his-tagged protein.
  • Ultimate Versatility Sartobind™ Lab IDA units are available with a 2.1 mL bed volume, providing binding capacities up to 7.5 mg his-tagged protein per unit.
  • Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications.
  • The units are compatible with most common chromatography buffers and can be regenerated for multiple purification cycles, ensuring high-cost efficiency.
  • Fewer preparative steps
  • Greatly reduced cycle times
  • Achieve higher yields in less time
  • Purify equipment-free or with an FPLC system
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 2.1 mL
Dead Volume 1.3 mL
Disposable Reusable
Filter Type Air Syringe Filter
For Use With (Equipment) Syringe
Material Polyester
Material (Membrane) Cellulose
Pore Size 5 μm
Sterility Non-sterile
For Use With (Application) His-tagged Protein Purification
Max. Pressure 0.6 mPa
Membrane Type Metal Chelate
Type Membrane Adsorber
Volume (Metric) 2.1 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.