Learn More
Description
- Enables clarification and filtration to be performed in a single step
- Designed for the clarification and sterile filtration of up to 50 mL mammalian cell culture
- Comprised of pouches of filter aid and vacuum filtration units
- For a single-step mammalian cell culture harvesting
- Gamma sterilized pouches
Specifications
Specifications
| Description | Cell Density: > 10 mio. cells/mL; Pore Size: 0.22 μm |
| Product Type | Filtration Kit |
| Quantity | 2 g diatomaceous earth as filter aid per unit of filtration |
| Includes | 1 x 180E01---------2 (24 x Sartolab RF 50, 0.22 μm, PES, single packed sterile), 2 x SDLKG-01.0------2 (48 x pouches of filter aid, 1 g), 24 filtration units |
| For Use With (Application) | Cell Culture Filtration |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.