Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Absorbent Pads

Catalog No. 5090117671 Shop All Sartorius Products
Change view
Click to view available options
Color:
Black
Dark Green
Gray
Green
White
Media Type:
Azide
Bismuth Sulfite
Caso
Cetrimide
Chapman
Chromocult
ECD
Endo
Glucose Tryptone
Jus de Tomate
MacConkey
Orange Serum
R2A
Sabouraud Agar
Schaufus Pottinger
Standard TTC
TGE
Tergitol TTC
VLB-S7-S
Wallerstein
m-FC
Pore Size:
0.45μm
0.65μm
1.2μm
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Media Type Pore Size
5090117671 Green MacConkey 0.45μm
5090117657 Dark Green TGE 0.45μm
5090117666 Black Chromocult 0.45μm
5090117638 Dark Green Azide 0.45μm
5090117642 Green Tergitol TTC 0.45μm
5090117647 Dark Green Caso 0.45μm
5090117653 Green Chapman 0.45μm
14-555-673 White Endo 0.45μm
14-555-677 Green Standard TTC 0.45μm
14-555-683 Gray Bismuth Sulfite 0.65μm
14-555-685 White VLB-S7-S 0.45μm
14-555-689 Gray Orange Serum 0.45μm
14-555-697 White Glucose Tryptone 0.45μm
14-555-702 White m-FC 0.45μm
14-555-704 Gray Sabouraud Agar 0.65μm
14-555-710 White Cetrimide 0.45μm
14-555-714 Green Jus de Tomate 0.45μm
14-555-722 Green R2A 0.45μm
5090117667 Green Wallerstein 0.45μm
5090117651 Green Schaufus Pottinger 1.2μm
5090117663 Green ECD 0.45μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 5090117671 Supplier Sartorius Supplier No. 1409750N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.

  • trouble-free storage up to 24 month
  • economical by time- and cost-saving
  • according to international standards and references
  • easy handling
  • validation guide available
  • consistent quality
  • highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color Green
Form Dehydrated
Manufacturing Origin Germany
Media Type MacConkey
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Enterobacteria, E. coli
Grid Color White
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.96 in.
Diameter (Metric) Outside 5 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.