Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ advance EDI Electodeionization Water System, ASTM Type 2
GSA_VA

Catalog No. 14556201 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
10 L/hr.
5 L/hr.
Mounting:
Benchtop
Wall
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Mounting
14-556-201 10 L/hr. Benchtop
14-556-200 10 L/hr. Wall
15-555-500 5 L/hr. Benchtop
15-555-501 5 L/hr. Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-556-201 Supplier Sartorius Supplier No. H2OEDI2T
Add to Cart
Add to Cart
Request a Quote

Outfit your lab with he Sartorisu™ arium™ advance EDI provide consistently high quality Type 2 water.

The arium™ advance EDI pure water system delivers Type II water with the best quality at rates of up to 10l/hr. The system attains the highest retention rates of ions along with optimal water yield, and reliably removes oxidants, heavy metal ions and particulates from feed water. The pure water produced is stored in arium™ bagtank systems. If a Sartopore™ 2 150 final filter is used in the system, the water is practically free of microorganisms.

  • Innovative iJust technology
  • Uses unique bagtank technology with dispense gun connections
  • User-friendly, easy operation
  • Unique pre-filter technology with activated carbon, phosphate-free catalyst and 5μm particle filter

Benefits:

  • Optimized water consumption
  • Meets customer's lab requirements and saves maintenance time
  • No user manual needed, intuitive operation of the menu
  • Highly efficient reduces colloids and chlorine, oxidizing agents, organic and water-soluble heavy metals

Applications:

  • Microbiological media and reagents
  • Solutions for chemical analysis and synthesis
  • Histology
  • ELISA, RIA
  • Buffers and pH solutions
  • Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Conductivity 0.2 to 0.07 Product water, Less than1500 feed water
Contaminant Rejection (Microbiological) Greater than99%
Contaminant Rejection (Organic) Greater than99%
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 10 L/hr.
Product Water Type Type 2
Remote Dispenser No
Resistivity <5 to 15 product water
TOC Monitor with UV Intensity Monitoring No
Total Organic Carbon <50 product water, <1000 feed water
Feedwater Temperature (English) 35.6°F to 86°F
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc
Humidity Range 80%
Mounting Benchtop
Particle Size <1/mL
pH Range 4 to 10
Storage Requirements 5°C to 45°C
Temperature Range (English) 35.6°F to 95°F
Temperature Range (Metric) 2°C to 35°C
Tolerance 4
Turbidity <1
Width (English) 13.7 in.
Width (Metric) 35 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 17.75 in.
Depth (Metric) 45.1 cm
Weight (English) 57.3 lb.
Weight (Metric) 26 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Daily Output 1 to 10 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type EDI Electrodeionization Water System
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.