Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium comfort II Combined System (TYPE 1 - Ultrapure Water and TYPE 2 - EDI Water)

Catalog No. 14556203
Click to view available options
Mounting:
Benchtop
Underbench
Wall

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

arium comfort II systems produce ASTM Type 1 ultrapure water and Type 2 pure water. The systems features electrodeionization (EDI) technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.

  • Flexible, space-saving twin technology
  • Efficient iJust technology
  • Uses disposable bagtank technology with dispense gun connections
  • Built-in TOC monitor analyzes and displays actual TOC values
  • Integrated UV lamp, 185/254nm, reduce the TOC ≤ 2ppb
  • Convenient SD card slot
  • User-friendly menu navigation

Benefits:

  • Produces Type 2 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
  • Optimized water consumption
  • Meet customer's lab requirements and saves maintenance time
  • Safe operation
  • Reliable TOC reduction
  • Supports reliable recording of important data
  • Easy operation
  • HPLC
  • AAS
  • Ion chromatography
  • Photometry
  • Microbiological media and reagents
  • Histology
  • ELISA, RIA
  • Buffers and pH solutions
  • Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.
TRUSTED_SUSTAINABILITY

Specifications

Conductivity 0.055 Product water, Less than1500 feed water
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate Up to 2 L/min.
Product Water Type Type 1 and 2
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Type Arium Comfort II Combined System
Feedwater Temperature (English) 35.6°F to 86°F
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc
Humidity Range 80%
Includes TOC monitor
Mounting Underbench
Particle Size <1/mL
pH Range 4 to 10
Storage Requirements 5°C to 45°C
Temperature Range (English) 35.6°F to 95°F
Temperature Range (Metric) 2°C to 35°C
Tolerance <4
Turbidity <1
Width (English) 17.12 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.74 in.
Depth (Metric) 47.6 cm
Weight (English) 79.36 lb.
Weight (Metric) 36 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.