Learn More
Sartorius arium™ Conductivity Meter for Deionization Cartridges
Description
The arium™ line also includes conductivity meters for checking the quality of product water. These convenient options are available in a choice of LED controlled display or digitally controlled display.
The LED controlled conductivity meter enables the user to monitor the conductivity of the product water with a red (>20μS/cm) or green (>20μS/cm) light. If the water does not meet the user's quality requirements, the LED will light up in red to alert the user.
The digitially controlled conductivity meter lets the user monitor the conductivity by individually defined setpoint limits (0.1–200μS/cm). The conductivity measurement range is between 0.056 and 200μS/cm.
Specifications
Specifications
Type | Conductivity Meter |
Display | Digital |
For Use With (Application) | Checking the quality of product water |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.