Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Balance and Scale Dust Covers

Click to view available options
For Use With (Equipment):
CPA Balance Models: CPA2202S, -4202S, -6202S, -620
CPA Balance Models: CPA223S, -323S, -423S, -623S,
CPA Balances, Except: CPA225D, -64, -124S, -224S
CPA Balances: CPA225D, -64, -124S, -224S, -324S
ED Series Models with Round Pan of 115 mm or 150 mm
ED Series Models with Square Pan
M-Power Balance Models: AZ1502, AZ1302, AZ3102, AZ
M-Power Balance Models: AZ212, AZ612
M-Power Series Balances: AZ64, AZ124, AZ214
Master Pro LA Balance
Type:
Cover, Dust, In-use
Dust Cover, Display Head Only
Dust Cover, Display Only
Dust Cover, Filter Weigher Display
Dust Cover, Toploader Display
One Piece Dust Cover
Description
Use with Sartorius balances.

Specifications
Specifications
For Use With (Equipment) | ED Series Models with Round Pan of 115 mm or 150 mm |
For Use With (Application) | For protecting a balance from dust and debris |
Type | Cover, Dust, In-use |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction