Learn More
Sartorius Connectors for Vivaflow™ Models
Catalog No. 14558326 Shop All Sartorius Products
Change view
Click to view available options
Shape:
Y
Catalog No. Shape
14-558-326 Y
1 options
Catalog No. 14-558-326 Supplier Sartorius Supplier No. VFA005
Connectors for Vivaflow 200
  • Y-Connector fits one Size 15 branch and splits into two size 16 branches
TRUSTED_SUSTAINABILITY
Specifications
Material PVC
Shape Y
Color Transparent
Product Type Y Connector
WARNING: Cancer - www.P65Warnings.ca.gov