Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Extra Thick Glass Fiber (GF) Pre-filters

Catalog No. 14555453
Click to view available options
Diameter (English):
1.96 in.
4.88 in.
5 in.
5.1 in.
Diameter (Metric):
5 cm
12.4 cm
12.7 cm
13 cm
Flow Rate:
348 to 490mL/min.
4.1 to 4.4 sec.
Grade:
13430
13440

Glass fiber prefilters, depth pre filters, for larger totally filterable volumes in clarification and sterile filtration.

The major use of all three glass fibre filters is as a depth prefilter, placed directly on top of a membrane filter, whereby the prefilter diameter specified for the holder must be chosen. Larger diameters would intrude under the sealing ring of the holder and cause leakage. The standard type 13400 contains an acrlic binder. It has a high particle loading capacity, but for very dirty liquids the thicker type 13430 can be more effective. 13440 ia a finer , binder-free type and is recommended for the prefiltration of relative clean solutions such as tissue culture media.Serial filtration may be necessary for difficult-to-filter liquids such as serum.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Filter Type Net Filter
Format Disc
For Use With (Equipment) Filtration Systems
Material Borosilicate Glass Microfiber
Particle Retention 0.7 μm
Sample Volume 120 mL
Sterility Non-sterile
Flow Rate 4.1 to 4.4 sec.
For Use With (Application) Prefiltration of clean solutions such as tissue culture media
Grade 13440
Hydrophobicity Hydrophilic
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.017 in.
Thickness (Metric) 0.44 mm
Type Glass Microfiber Filter
Diameter (English) 1.96 in.
Diameter (Metric) 5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.