Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Gridded Nonsterile Cellulose Nitrate Membrane Filters: 0.45μm Pore Size

Catalog No. 5090117782 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
50 mm
Grid Color:
Black
Dark Green
Green
White
Color:
Gray
Green
White
Quantity:
100 Pk.
1000 Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Grid Color Color Quantity
5090117782 50 mm Green White 1000 Pk.
5090117761 47 mm Green White 1000 Pk.
5090117763 50 mm White Gray 100 Pk.
5090117774 47 mm Dark Green Green 1000 Pk.
5090117776 50 mm Dark Green Green 100 Pk.
5090117777 50 mm Dark Green Green 1000 Pk.
14-555-594 25 mm Black White 100 Pk.
14-555-595 47 mm Black White 100 Pk.
14-555-623 47 mm White Gray 100 Pk.
14-555-622 25 mm White Gray 100 Pk.
14-555-653 47 mm Green White 100 Pk.
5090117781 50 mm Green White 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 5090117782 Supplier Sartorius Supplier No. 1390650R
Add to Cart
Add to Cart

The Nonsterile Membrane Filters are made of cellulose nitrate (cellulose mixed ester), cellulose acetate or regenerated cellulose. They can be used for colony counting, particle testing and microscopy. Choose between gridded or non-gridded, membranes, either with or without hydrophobic edge.

If agar plates or absorbent pads to be wetted with liquid culture medium are used instead of Nutrient Pad Sets, Sartorius cellulose nitrate (cellulose ester) membrane filter are recommended. Naturally, the membrane filters must be free of microbes. For this purpose, they can be boiled or autoclaved. These membranes are offered in a choice of three different colors to suit the specific test application, and provide a high-contrast background to colonies or particles.These membrane filters are in accordance with the following norms: ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780. In addition to this they have been manufactured for use especially at the same time with Sartorius Nutrient Pad Sets in accordance with many international regulations and recommendations: international pharmacopoeias, DIN and ISO Standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA. All membranes are made of cellulose nitrate (cellulose ester), a material which assures effective retention with high flow rates and optimum colony growth. The printed grid divides the filtration area into 130 squares, each measuring 3.1 x 3.1 mm makes the counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth.

  • Outstanding recovery rates for microorganisms
  • 0.45 μm are acc. to ISO 7704
  • Three different colors available
  • Design: 25, 47 or 50 mm in diameter, white, grey or green and gridded
  • Growth Promotion Test acc. to ISO 7704: no enhancement or inhibition by the grid lines, no enhancement or inhibition due to chemical extractables
TRUSTED_SUSTAINABILITY

Specifications

Color White
Disposable Single-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Nitrate
Material (Membrane) Cellulose Nitrate
Pore Size 0.45 μm
Sterility Non-sterile
For Use With (Application) For Colony Count, Sterility Testing, Particle Count, Microscopy
Grid Color Green
Quantity 1000 Pk.
Type Membrane Filter
Diameter (Metric) 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.