Description
The innovative weigh cell technology, the rugged construction of the balance housing, the sleek stainless steel weighing pan and the keypad sealed by a membrane overlay assure high dependability and accuracy, even during frequent use. Talent series balances offer various application programs as standard features to make routine work easy: weighing in percent, net-total formulation, weigh averaging / dynamic weighing, counting of small parts and mass unit conversion by toggling between two weight units. Each model comes standard with a bidirectional RS-232C interface port. This means no extra cost if you need to log the balance-generated results on an optional printer or connect a remote display for use in the educational sector.
- High-contrast LCD
- All-glass draft shield chamber
- Bidirectional RS-232C interface port
- Small Footprint Space Saver
- Built-in overload protection
- 4 digital filter levels to adapt the balance to the conditions at the place of use
Specifications
Specifications
| Readability | 0.1 mg |
| Material | Durable ABS Housing |
| Model | AZ124 |
| Stabilization Time | 3 sec. |
| Type | Analytical Balance |