Learn More
Sartorius M-Power™ Analytical Balances
Catalog No. p-4183226
Click to view available options
Model:
AZ124
AZ214
AZ64
The balance for first-time users at vocational and professional schools and universities
The innovative weigh cell technology, the rugged construction of the balance housing, the sleek stainless steel weighing pan and the keypad sealed by a membrane overlay assure high dependability and accuracy, even during frequent use. Talent series balances offer various application programs as standard features to make routine work easy: weighing in percent, net-total formulation, weigh averaging / dynamic weighing, counting of small parts and mass unit conversion by toggling between two weight units. Each model comes standard with a bidirectional RS-232C interface port. This means no extra cost if you need to log the balance-generated results on an optional printer or connect a remote display for use in the educational sector.
  • High-contrast LCD
  • All-glass draft shield chamber
  • Bidirectional RS-232C interface port
  • Small Footprint Space Saver
  • Built-in overload protection
  • 4 digital filter levels to adapt the balance to the conditions at the place of use
TRUSTED_SUSTAINABILITY
Specifications
Readability 0.1 mg
Material Durable ABS Housing
Model AZ64
Stabilization Time 3 sec.
Type Analytical Balance
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.