Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Minisart™ HY Syringe Filters

Catalog No. 14555305 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.2 μm
Sterility:
Non-sterile
Sterile
Hold up Volume (After Air Purge):
0.15 mL
0.25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Sterility Hold up Volume (After Air Purge)
14-555-305 0.2 μm Non-sterile 0.15 mL
14-555-283 0.2 μm Sterile 0.25 mL
14-555-284 0.2 μm Non-sterile 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-555-305 Supplier Sartorius Supplier No. 17595HYQ

Designed for fast and reliable sterile venting and gas filtration. Sartorius Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0μm pore sizes. Sterile option available.

Fast and reliable sterile venting and filtration
  • Clean, safe filters that are almost free of leachables and extractables
  • Reliably removes particles and microorganisms with no leakage
  • Has minimal effect on the ingredients of the filtered solution
  • Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
  • Wide variety of lab uses:
    • Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
    • Sterile venting of small fermenters or bioreactors and of containers for culture media
    • Also used for aqueous and oily ear- or eye-solutions and other drugs
Convenient and economical
  • Complete units are ready to use
  • Single use/disposable filter units
  • Proven quality and 100% tested during manufacture
  • Sterile (via ethylene oxide) and non-sterile units available
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Single-use
Material PTFE
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 5.16 cm2
Hold up Volume (After Air Purge) 0.15 mL
Housing Material Medical acrylic (MBS)
Hydrophobicity Hydrophobic
Inlet Style Female Luer Lock
Outlet Style Male Luer Lock
Pressure Rating 4.5 bar
Quantity 500/Pk.
Tip Material Cryolite
Type Syringe Filter
Diameter (Metric) 26 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.