Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Nutrient Pad Sets (NPS) 47mm diameter

Catalog No. 5090117668 Shop All Sartorius Products
Change view
Click to view available options
Color:
Gray
Green
White
Grid Color:
Black
Dark Green
Green
White
Pore Size:
0.45μm
0.65μm
0.8μm
Media Type:
Acid-tolerant
Azide
Bismuth Sulfite
Caso
Cetrimide
Chapman
Chromocult
Endo
Glucose Tryptone
Jus de Tomate
MacConkey
Malt Extract
Orange Serum
R2A
Standard
Standard TTC
TGE
Tergitol TTC
VLB-S7-S
Wallerstein
Yeast Extract
m-FC
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Grid Color Pore Size Media Type
5090117668 Green Green 0.45μm Yeast Extract
14-555-669 Green Dark Green 0.45μm Azide
14-555-670 White Green 0.45μm Endo
14-555-674 Green Dark Green 0.45μm Standard TTC
14-555-678 White Green 0.45μm Tergitol TTC
14-555-681 Gray White 0.65μm Bismuth Sulfite
14-555-684 White Green 0.45μm VLB-S7-S
14-555-688 Green Dark Green 0.45μm Orange Serum
14-555-690 Green Dark Green 0.45μm Caso
14-555-691 Green Dark Green 0.45μm Standard
14-555-696 White Green 0.45μm Glucose Tryptone
14-555-700 White Green 0.45μm m-FC
14-555-709 White Green 0.45μm Cetrimide
14-555-708 White Green 0.45μm Chapman
14-555-711 Green Dark Green 0.45μm TGE
14-555-713 Green Dark Green 0.45μm Jus de Tomate
14-555-721 Green Dark Green 0.45μm R2A
14-555-723 Gray White 0.8μm Malt Extract
14-555-724 White Black 0.45μm Malt Extract
14-555-728 White Green 0.45μm Acid-tolerant
14-555-726 Green Dark Green 0.45μm Wallerstein
14-555-725 Gray White 0.45μm Chromocult
14-555-729 Gray White 0.45μm MacConkey
14-555-727 Gray White 0.8μm Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 5090117668 Supplier Sartorius Supplier No. 1409047N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.

  • trouble-free storage up to 24 month
  • economical by time- and cost-saving
  • according to international standards and references
  • easy handling
  • validation guide available
  • consistent quality
  • highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 6222
Color Green
Form Dehydrated
Manufacturing Origin Germany
Media Type Yeast Extract
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Total colony count
Grid Color Green
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.85 in.
Diameter (Metric) Outside 4.7 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.