Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Nutrient Pad Sets (NPS) for Microsart™ e.motion Dispenser

Catalog No. 5090117656 Shop All Sartorius Products
Change view
Click to view available options
Media Type:
Cetrimide
Endo
Orange Serum
Schaufus Pottinger (m-Green Yeast and Mold)
Standard TTC
TGE
Tergitol TTC
Wort
Diameter (Metric) Outside:
4.7 cm
5 cm
Pore Size:
0.45μm
0.65μm
0.8μm
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Diameter (Metric) Outside Pore Size
5090117656 TGE 4.7 cm 0.45μm
5090117644 Wort 5 cm 0.65μm
5090117639 Endo 5 cm 0.45μm
5090117645 Orange Serum 4.7 cm 0.45μm
5090117641 Tergitol TTC 4.7 cm 0.45μm
5090117646 Orange Serum 5 cm 0.45μm
5090117654 Cetrimide 4.7 cm 0.45μm
5090117655 Cetrimide 5 cm 0.45μm
5090117661 Schaufus Pottinger (m-Green Yeast and Mold) 5 cm 0.8μm
14-555-672 Endo 4.7 cm 0.45μm
14-555-676 Standard TTC 4.7 cm 0.45μm
14-555-682 Wort 4.7 cm 0.65μm
14-555-712 TGE 5 cm 0.45μm
5090117643 Tergitol TTC 5 cm 0.45μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 5090117656 Supplier Sartorius Supplier No. 1407647RDN
Add to Cart
Add to Cart

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are sterile, dehydrated culture media designed for use with membrane filter methods. Each Nutrient Pad Set includes the appropriate membrane filters (in 47 or 50mm diameters), which are also sterile and individually packaged. Just moisten the Nutrient Pads with sterile and demineralized (or distilled) water and they are immediately ready to use. They comply with international regulations and recommendations including, but not limited to: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA.

  • Trouble-free storage up to 24 months
  • Economical -- time and cost savings
  • Easy handling
  • Validation guide available
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color Green
Form Dehydrated
Manufacturing Origin Germany
Media Type TGE
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Total colony count
Grid Color Dark Green
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.85 in.
Diameter (Metric) Outside 4.7 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.