Learn More
Sartorius Sterility Testing Reusable System: Tubing Clamp
Click to view available options
:
Tubing Clamp; Sterile: No
Description
- Clamps off tubing between T or X adapter and cap
- Use while culture medium is filled in a filter holder that is not clamped off
- Not autoclavable
Specifications
Specifications
Autoclavable | Not Autoclavable |
Disposable | Reusable |
For Use With (Equipment) | Reusable Sterility Testing System |
Material | Stainless Steel |
Sterility | Non-sterile |
Type | Tube Clamps |