Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Vivapore™ 5 Static Concentrators
Click to view available options
Quantity:
100/Pk.
30/Pk.
Description
Economic and user-friendly
- No need for additional equipment, pressure or a vacuum source
- Individual devices with disposable plastic stands for up to four units
- Simple concentration technique:
- Just fill the unit with the solution to be concentrated
- Wait for the desired concentration level to be achieved
- Then pipet the concentrated sample from the dead-stop reservoir
- Separate concentration chambers eliminate the risk of cross-contamination
- Each unit features a high-flow, 7.5 kDa PES membrane
- Ideal for samples that are sensitive to shearing forces or to pressure-induced stress
- Well suited for samples that require processing in a cold room environment
- Perfect for a variety of clinical and general research applications
- Urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis
- Concentrating macromolecules in cerebrospinal fluid (CSF) or other biological solutions

Specifications
Specifications
Disposable | Single-use |
Filter Type | Ultrafiltration |
Material (Membrane) | PES |
MWCO | 7.5 kDa |
Sample Volume | 5 mL |
Sterility | Non-sterile |
Filtration Area | 20 cm2 |
Housing Material | Styrene acrylonitrile (SAN) |
Quantity | 100/Pk. |
Type | Static Concentrator |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction