Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane
GSA_VA

Catalog No. 14558417
Click to view available options
MWCO:
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.

Twin vertical membranes and optimal design for fast, safe concentration

  • Patented twin vertical membrane design for ultra-fast processing speeds
  • In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5-30 minutes
  • Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
  • Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
  • High-flux PES membranes feature very low protein binding and low extractables

Designed for maximum flexibility and sample recovery

  • Low internal surface and membrane area for superior recoveries from very dilute solutions
  • Two methods for easy concentrate recovery:
    • Directly pipet the concentrate from the dead stop pocket in the concentrator
    • Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
    • Both methods provide near-total concentrate recovery of greater than 90%
  • Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
  • Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 10 kDa
Sample Volume Up to 3 mL
Sterility Non-sterile
Filtration Area 1.3 cm2
For Use With (Application) Concentration, desalting and buffer applications
Housing Material Polycarbonate
Max. Force Swing bucket rotor: 4000 x g, Fixed angle rotor: 9000 x g (3 to 50 kDa MWCO); 12,000 x g (100 kDa+ MWCO)
Quantity 25/Pk.
Type Centrifugal Concentrator
Volume (Metric) 2 mL
Length (Metric) 126 mm
Width (Metric) 17.8 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.