Learn More
SAS-6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301801A
Description
The recommended shelf life for this product is 1 year from date of receipt.
SAS6 is necessary for centrosome duplication and functions during procentriole formation; SAS6 functions to ensure that each centriole seeds the formation of a single procentriole per cell cycle Strnad et al., (2007) [PubMed 17681132].Specifications
| SAS-6 | |
| Polyclonal | |
| Unconjugated | |
| SASS6 | |
| SAS-6, SAS6, hsSAS-6, spindle assembly abnormal protein 6 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 163786 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q6UVJ0 | |
| SASS6 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.