Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAS™ Legionella Test
Click to view available options
Quantity:
2 x1 mL
Description
Includes
Test devices; Disposable specimen droppers
Requires
Timer; Specimen Collection Container
- CLIA Complexity: Non-waived
- CPT Code: 87449
- Sensitivity: 90.5%
- Specificity: 95%
- Rapid membrane-based immunoassay for the qualitative detection of Legionella pneumophila serogroup 1 antigens in urine
- Results within 15 minutes
Specifications
Specifications
Content And Storage | Room Temperature |
CPT Code | 87449 |
Clia Complexity | Non-waived |
Detectable Analytes | Legionella |
Sensitivity | 91% |
Type | Legionella Control |
Includes | Test device, disposable specimen droppers, 1mL positive control and 1mL negative control |
Quantity | 2 x1 mL |
Specificity | 0.95 |
Test Time | >15 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction