Learn More
SAS10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302067AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
Essential for gene silencing: has a role in the structure of silenced chromatin. Plays a role in the developing brain.Specifications
SAS10 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NQZ2 | |
UTP3 | |
Between 429 and 479 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UTP3 | |
Something about silencing protein 10, Charged amino acid-rich leucine zipper 1, CRL1, Disrupter of silencing SAS10, UTP3 homolog, UTP3, CRLZ1, SAS10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57050 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.