Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SASH1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126650V
Description
SASH1 Polyclonal antibody specifically detects SASH1 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SASH1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
dJ323M4, dJ323M4.1, KIAA0790RP3-323M4.1, PEPE1, Proline-glutamate repeat-containing protein, SAM and SH3 domain containing 1,2500002E12Rik, SAM and SH3 domain-containing protein 1, SH3D6A | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human SAM and SH3 domain containing 1 (NP_056093.3). | |
0.1 mL | |
Cell Cycle and Replication | |
23328 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction