Learn More
Invitrogen™ SASH1 Monoclonal Antibody (X1)

Description
Peptide Sequence: ENTSLQEHGV KLGPALTRKV SCARGVDLET LTENKLHAEG IDLTEEPYSD KHGRCGIPEA LVQRYAEDLD QPERDVAANM DQIRVKQLRK QHRMAIPSGG LTEICRKPVS.
Specifications
Specifications
| Antigen | SASH1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | X1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SASH1 |
| Gene Accession No. | O94885 |
| Gene Alias | 1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.