Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SASH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550749 Supplier Invitrogen™ Supplier No. PA550749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SASH1 protein.

May have a role in a signaling pathway. Could act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SASH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SASH1
Gene Accession No. O94885, P59808
Gene Alias 1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A
Gene Symbols SASH1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human SAM and SH3 domain containing 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23328, 70097
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.