Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SASH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590318 Supplier Invitrogen™ Supplier No. PA590318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEDAGAAGPG PEPEPEPEPE PEPAPEPEPE PKPGAGTSEA FSRLWTDVMG ILDGSLGNID DLAQQYADYY NTCFSDVCER MEELRKRRVS QDLEVEKPDA SPTSLQLRSQ IEESLGFCSA VSTPEVERKN PLHKSNSEDS SVGKGDWKKK NKYFWQNFRK NQKGIMRQTS KGEDVGYVAS EITMSDEERI QLMMMVKEKM ITIEEALARL KEYEAQHRQS AALDPADWPD GSYPTFDGSS NCNSREQSDD ETEESVKFKR LHKLVNSTRR VRKKLIRVEE MKKPSTEGGE EHVFENSPVL; Positive Samples: U-87MG, Mouse lung.

May have a role in a signaling pathway. Could act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SASH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SASH1
Gene Accession No. O94885, P59808
Gene Alias 1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A
Gene Symbols SASH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SASH1 (NP_0560933).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23328, 70097
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.