Learn More
SASH1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00646T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Northern white-cheeked gibbon
May have a role in a signaling pathway. Could act as a tumor suppressor.Specifications
SASH1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O94885 | |
SASH1 | |
Between 750 and 800 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
SASH1 | |
SAM and SH3 domain-containing protein 1, Proline-glutamate repeat-containing protein, SASH1, KIAA0790, PEPE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23328 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.