Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SASH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551511 Supplier Invitrogen™ Supplier No. PA551511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKTLHELLER IGLEEHTSTL LLNGYQTLED FKELRETHLN ELNIMDPQHR AKLLTAAELL LDYDTGSEEA EEGAESSQEP VAHTVSEPKV DIPRDSGCFE GSESGRDDAE LAGTEEQLQG LSL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

The protein encoded by this gene contains a Src homology-3 (SH3) domain and a sterile alpha motif (SAM), both of which are found in proteins involved in cell signaling. This protein may function as a signaling adapter protein in lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SASH3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SASH3
Gene Accession No. O75995
Gene Alias 1200013B08Rik; 753P9; AW413946; CXorf9; HACS2; HXCXORF9; Novel SH3 and SAM domain containing protein; RGD1560293; SAM and SH3 domain containing 3; SAM and SH3 domain-containing protein 3; SASH3; SH3 protein expressed in lymphocytes; SH3 protein expressed in lymphocytes homolog; SH3D6C; Sly; SLY1
Gene Symbols SASH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SASH3. Recombinant protein control fragment (Product #RP-91940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.