Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SASS6 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093343H
Description
SASS6 Polyclonal antibody specifically detects SASS6 in Human samples. It is validated for Western BlotSpecifications
SASS6 | |
Polyclonal | |
Western Blot | |
DKFZp761A078, FLJ22097, HsSAS-6, MGC119440, SAS-6, SAS6hsSAS-6, spindle assembly 6 homolog (C. elegans), spindle assembly abnormal protein 6 homolog | |
The immunogen recognized by this antibody maps to a region between residue 607 and 657 of human spindle assembly 6 using the numbering given in entry NP_919268.1 (GeneID 163786). | |
0.1 mL | |
Cell Cycle and Replication | |
163786 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction